Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PPBP (Mouse) ELISA Kit

Catalog No. 89100429 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-429 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-429 Supplier Abnova™ Supplier No. KA2149

missing translation for 'validMainframeMsg'

Includes

  • TCK-1 Microplate (Item A): coated with anti-Mouse TCK-1
  • Wash Buffer Concentrate (20x) (Item B)
  • Standards (Item C): recombinant Mouse TCK-1
  • Assay Diluent C (Item L): diluent buffer. For Standard/Sample (serum/plasma samples/cell culture medium) diluent
  • Assay Diluent B (Item E): 5x concentrated buffer. For detection antibody and HRP Streptavidin diluent
  • Detection Antibody TCK-1 (Item F): biotinylated anti-Mouse TCK-1
  • HRP-Streptavidin concentrate (Item G): 600x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H)
  • Stop Solution (Item I): 0.2M sulfuric acid

Use to measure quantitatively mouse Ppbp. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for TCK-1 coated on a 96-well plate. Standards and samples are pipetted into the wells andTCK-1 present in a sample is bound to the wells by the immobilized antibody

Specifications

Conjugate HRP
Cross Reactivity Mouse
Description Ppbp (Mouse) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of mouse Ppbp.
Detection Limit 14pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma, Cell Culture Supernatant
Sample Volume 100 μL
Sensitivity <14 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Ppbp (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.