Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PPBP (Rat) ELISA Kit

Catalog No. 89100430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-430 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-430 Supplier Abnova™ Supplier No. KA2150

Includes

  • TCK-1 Microplate (Item A): 96 wells coated with anti-Rat TCK-1
  • Wash Buffer Concentrate (20x) (Item B)
  • Standards (Item C): 2 vials, recombinant Rat TCK-1
  • Assay Diluent (Item E): 15mL of 5x concentrated buffer. For Standard/Sample (serum/plasma samples/cell
  • culture medium) diluent
  • Detection Antibody TCK-1 (Item F): 2 vial of biotinylated anti-Rat TCK-1
  • HRP-Streptavidin concentrate (Item G): 8μl 20,000x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H): 12mL of 3,3',5,5'-tetramethylbenzidine (TMB) in buffered solution
  • Stop Solution (Item I): 2M sulfuric acid

Use to measure quantitatively rat Ppbp. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for Rat TCK-1 coated on a 96-well plate

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Ppbp (Rat) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of rat Ppbp.
Detection Limit 80pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma, Cell Culture Supernatant
Sample Volume 100 μL
Sensitivity 80 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Ppbp (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.