Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Pregnanediol-3-Glucuronide ELISA Kit

Catalog No. 89100935 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-935 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-935 Supplier Abnova™ Supplier No. KA3398
Add to Cart
Add to Cart

Includes

  • Coated Clear 96 Well Plates: Clear 1 by 8 break-apart strip well plastic microtiter plate coated with goat anti-rabbit IgG, 1 plate
  • Pregnanediol-3-Glucuronide (PDG) Standard: Pregnanediol-3-Glucuronide (PDG) at 500ng/mL in a special stabilizing solution
  • Pregnanediol-3-Glucuronide (PDG) Antibody
  • Pregnanediol-3-Glucuronide (PDG) Conjugate: Pregnanediol-3-Glucuronide-peroxidase conjugate in a special stabilizing solution
  • Assay Buffer Concentrate: A 5X concentrate
  • Wash Buffer Concentrate: A 20X concentrate
  • TMB Substrate
  • Stop Solution: A 1M solution of hydrochloric acid. CAUSTIC
  • Plate Sealer

Use to detect quantitatively of pregnanediol-3-glucuronide in urine and fecal samples, or in extracted serum and plasma. Enzyme-linked immunosorbent assay.

The Pregnanediol-3-Glucuronide ELISA Kit uses a specifically generated antibody to measure PDG and its metabolites in urine and fecal samples, or in extracted serum and plasma

Specifications

Calibration Range 0.391 to 50ng/mL
Conjugate HRP
Description Pregnanediol-3-Glucuronide ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of pregnanediol-3-glucuronide in urine and fecal samples, or in extracted serum and plasma.
Detection Limit 0.320ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Dried Fecal Extract, Extracted Serum and Plasma, Tissue Culture Medium, Urine
Sample Volume 50 μL
Sensitivity 0.180 ng/mL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Pregnanediol-3-Glucuronide ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.