Learn More
Description
- Sample Type: Serum
- Sample Volume: 50μL
Quantification
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Conjugate | HRP |
| Description | Pregnenolone ELISA Kit is an enzyme immunoassay for the quantitative measurement of pregnenolone in serum. |
| Detection Limit | 0.05ng/mL |
| Detection Method | Colorimetric |
| Includes | 96-well Antibody coated microwell, 0.3mL HRP Conjugate concentrate, 6 vials Pregnenolone standard, 0.5mL Control, 50mL Wash Buffer 10X concentrate, 15mL Assay Buffer, 16mL TMB Substrate, 6mL stopping solution |
| Quantity | 1 Kit |
| Sample Type | Serum |
| Sample Volume | 50 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
