Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Progesterone ELISA Kit

Catalog No. 89100170 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-170 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-170 Supplier Abnova™ Supplier No. KA0299
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Goat anti-Mouse IgG Plate, 96 well
  • Progesterone Alkaline
  • Phosphatase Conjugate, 6mL
  • Progesterone Monoclonal Antibody, 6mL
  • Assay Buffer, 30mL
  • Wash Buffer Concentrate, 30mL
  • Progesterone Standard, 0.5mL
  • Steroid Displacement
  • Reagent, 1mL
  • pNpp Substrate, 20mL
  • Stop Solution, 5mL
  • Plate Sealer, 1 slice

Use to measure quantitatively Progesterone in biological fluids. A competitive immunoassay.

Progesterone ELISA kit is a competitive immunoassay for quantitative determination of Progesterone in biological fluids

Specifications

Calibration Range 15.62 to 500pg/mL
Conjugate AP
Description Progesterone ELISA Kit is a competitive immunoassay for the quantitative determination of Progesterone in biological fluids.
Detection Limit 8.57pg/mL
Detection Method Colorimetric
Includes Microwells (96 wells), EIA conjugate (5mL), EIA antibody (5mL), Assay buffer (27mL), Wash buffer concentrate (27mL), Standard (0.5mL), pNpp substrate (20mL), Stop solution (5mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Biological fluid
Sample Volume 100 μL
Sensitivity 8.57 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
Form Lyophilized
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.