Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Progesterone (Human) ELISA Kit

Catalog No. 89100016 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-016 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-016 Supplier Abnova™ Supplier No. KA0235

missing translation for 'validMainframeMsg'

Includes

  • Goat Anti-Rabbit IgG-coated microtiter wells, 96 wells
  • Progesterone Reference Standards: 0, 0.5, 3.0, 10, 25, and 50ng/mL. Liquids, ready to use, 0.5mL each
  • Rabbit Anti-Progesterone Reagent (pink color), 7mL
  • Progesterone-HRP Conjugate Concentrate (11x), 1.3mL
  • Progesterone-HRP Conjugate Diluent, 13mL
  • Progesterone Control 1, Liquid, Ready to use, 0.5mL
  • Progesterone Control 2, Liquid, Ready to use, 0.5mL
  • TMB Reagent (One-Step), 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively progesterone. Sandwich enzyme immunoassay.

The progesterone EIA is based on the principle of competitive binding between progesterone in the test specimen and progesterone-HRP conjugate for a constant amount of rabbit anti-progesterone

Specifications

Calibration Range 0 to 50ng/mL
Conjugate HRP
Cross Reactivity Human
Description Progesterone (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of progesterone.
Detection Limit 0.3ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 25 μL
Sensitivity 0.0625 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.