Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Prolactin ELISA Kit

Catalog No. 89100937 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-937 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-937 Supplier Abnova™ Supplier No. KA3400
Add to Cart
Add to Cart

Includes

  • Clear Coated 96 Well Plate: Clear plastic microplate with break-apart strips coated with mouse anti-Prolactin, 1 plate
  • Prolactin Standard: Two vials of recombinant human Prolactin at 500pg. Store at -20°C, 2 Vials
  • Prolactin Conjugate: A polyclonal antibody to Prolactin labeled with peroxidase. Store at -20°C, 5mL
  • Assay Buffer Concentrate: A 5X concentrate, 28mL
  • Wash Buffer Concentrate: A 20X concentrate, 30mL
  • TMB Substrate, 11mL
  • Stop Solution: A 1N hydrochloric acid solution. Caustic, 5mL
  • Plate Sealer, 2 unit

Use to detect quantitatively of prolactin in biological samples and tissue culture media. Enzyme-linked immunosorbent assay.

The kit has been tested to measure human and elephant serum prolactin

Specifications

Calibration Range 15.625 to 1000pg/mL
Conjugate HRP
Description Prolactin ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of prolactin in biological samples and tissue culture media.
Detection Limit 13.4pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum, Tissue Culture Medium
Sample Volume 50 μL
Sensitivity 11.7 pg/mL
Content And Storage Store the standard and conjugate at -20°C. Store all other components at 4°C. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type Prolactin ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.