Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Prostacyclin ELISA Kit

Catalog No. 89100167 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-167 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-167 Supplier Abnova™ Supplier No. KA0304
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One 96 Well Donkey anti-Sheep IgG Plate
  • Urinary Prostacyclin Conjugate: A blue solution of alkaline phosphatase conjugated with 6-keto-PGF1α
  • Urinary Prostacyclin Antibody: A yellow solution of a sheep polyclonal antibody to 6k-PGF1α and 2,3d-6k-PGF1α
  • Assay Buffer: Tris buffered saline containing proteins and detergents and sodium azide as preservative
  • Wash Buffer Concentrate
  • Urinary Prostacyclin Standard: A solution of 100,000pg/mL 2,3d-6k-PGF1α
  • p-Npp Substrate
  • Stop Solution: A solution of trisodium phosphate in water. Keep tightly capped. Caution: Caustic
  • Plate Sealer

Use to measure quantitatively Prostacyclin (PGI2) breakdown products in urine. A competitive immunoassay.

The Prostacyclin ELISA kit is a competitive immunoassay for the quantitative determination of Prostacyclin (PGI2) breakdown products in urine

Specifications

Calibration Range 7.81 to 2000pg/mL
Conjugate AP
Description Urinary Prostacyclin ELISA Kit is a competitive immunoassay for the quantitative determination of Prostacyclin (PGI2) breakdown products in urine.
Detection Limit 6.58pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Urine
Sample Volume 100 μL
Sensitivity 6.58 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.