Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PTH (Human) ELISA Kit

Catalog No. 89024465 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-465 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-465 Supplier Abnova™ Supplier No. KA0924
Add to Cart
Add to Cart

For the quantitative measurement of intact-PTH (Parathyroid Hormone) in Human Serum

Specifications

Calibration Range 0 to 210pg/mL
Conjugate Biotin
Cross Reactivity Human
Description PTH (Human) ELISA Kit is used for the quantitative measurement of intact-PTH (Parathyroid Hormone) in human serum.
Detection Method Colorimetric
Includes Microwell coated with Sterptavidin 12 x 8 x 1, 7mL Biotinylated PTH Antibody (reagent 1), Peroxidase (Enzyme) labeled 7mL PTH Antibody (reagent 2), 20mL TMB Substrate (reagent B), Diluent (equine Serum) for patient samples read off-scale (reagent 3), 30mL ELISA Wash concentrate (Saline with Surfactant) (reagent A), 20mL stop solution, 5mL Reconstitution solution containing surfactant (reagent 4), calibrator (A,B,C,D,E,F) (CAL) 0.5mL, 0.5mL Controls 1 and 2 (CTRL)
Quantity 96 Assays
Sample Type Serum
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type PTH (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.