Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PTX3 (Human) ELISA Kit

Catalog No. 89101028 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma (EDTA), Saliva, Serum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-028 CCS, Plasma (EDTA), Saliva, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-028 Supplier Abnova™ Supplier No. KA3349
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human PTX3

Specifications

Calibration Range 312 to 20000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description PTX3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human PTX3.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-human PTX3 antibody, Lyophilized human PTX3 standards (20ng x 2), Biotin conjugated anti-human PTX3 antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated) (130uL), ABC diluent buffer (12mL),TMB color developing agent (10mL), TMB Stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma (EDTA), Saliva, Serum
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type PTX3 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.