Learn More
Specifications
Specifications
| Calibration Range | 312 to 20,000 pg/mL |
| Content And Storage | Store the kit at -20°C |
| Description | RBP4 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human RBP4. |
| Conjugate | Biotin |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Product Type | RBP4 (Human) ELISA kit |
| Sample Type | Body Fluids, Cell Culture Supernatant, Plasma, Serum, Tissue Lysates |
| Sufficient For | 96 Tests |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.