Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rbp4 (Mouse) ELISA Kit

Catalog No. 89028740 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-740 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-101-128 CCS, Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-740 Supplier Abnova™ Supplier No. KA2008
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Rbp4

Specifications

Calibration Range 0.132 to 20ng/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Rbp4 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Rbp4.
Detection Limit <10pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 10ng x 2 vials lyophilized recombinant Mouse Rbp4 standard, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Rbp4 Antibody, 12mL Antibody Diluent Buffer, 130μL Avidin -Biotin
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Rbp4 (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.