Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rbp4 (Mouse) ELISA Kit

Catalog No. 89101128 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
CCS, Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-128 CCS, Plasma, Serum
89-028-740 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-128 Supplier Abnova™ Supplier No. KA3832
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Rbp4

Specifications

Conjugate Biotin
Cross Reactivity Mouse
Description Rbp4 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Rbp4.
Detection Limit 6ng/mL
Detection Method Colorimetric
Includes Mouse RBP4 Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against mouse RBP4, Sealing Tapes: Each kit contains 3 precut, pressure sensitive sealing tapes, which can be cut to fit the format of the individual assay (3), Mouse RBP4 Standard: Mouse RBP4 in a buffered protein base (lyophilized) (20ng), Biotinylated Mouse RBP4 Antibody (70x): A 70-fold concentrated biotinylated polyclonal antibody against mouse RBP4 (105μL), MIX Diluent Concentrate (10x): A 10-fold concentrated buffered protein base (30mL), Wash Buffer Concentrate (20x): A 20-fold concentrated buffered surfactant (30mL x 2), Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate (80μL), Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine (8mL), Stop Solution: A 0.5 N hydrochloric acid to stop the chromogen substrate reaction (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
For Use With (Application) Quantification
Product Type Rbp4 (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.