Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RealBlue Peroxidase Substrate Kit

Catalog No. 89027129 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-129 500 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-129 Supplier Abnova™ Supplier No. KA0565
Add to Cart
Add to Cart

For detection of least-abundant markers

50-100 times more sensitive than DAB and is non-carcinogenic. RealBlue forms a blue chromogenic product, providing excellent contrast with DAB and other substrates for multiple labeling experiments.

Immunohistochemistry

Specifications

Description RealBlue Peroxidase Substrate Kit is 50-100 times more sensitive than DAB and is non-carcinogenic. Strongly recommended for the detection of least-abundant markers. RealBlue forms a blue chromogenic product, providing excellent contrast with DAB and other substrates for multiple labeling experiments.
Product Type RealBlue Peroxidase Substrate kit
Quantity 500 Slides
For Use With (Application) Immunohistochemistry (IHC)
Content And Storage Room Temperature
Kit Contents 100mL Realblue Peroxidase Substrate, 10mL (10X) Blocking solution concentrate, 50mL Orcein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.