Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RED Phosphatase Substrate Kit

Catalog No. 89027130 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1000 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-130 1000 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-130 Supplier Abnova™ Supplier No. KA0566

Immunohistochemistry

Specifications

Description RED Phosphatase Substrate Kit contains reagents for the detection of alkaline phosphatase-labeled conjugates. RED reagents form a brilliant scarlet reaction product that is stable in organic solvents. Sections reacted with RED reagents can be dehydrated through graded alcohols and xylene, then mounted in xylene-based mounting media for permanent documentation. The hematoxylin counterstain, Contrast BLUE, provides sharp contrast and excelllent nuclear detail.
Product Type RED Phosphatase Substrate kit
Quantity 1000 Slides
For Use With (Application) Immunohistochemistry (IHC)
Content And Storage Store the kit at 4°C
Kit Contents 10mL Activator solution, 10mL RED solution, 50mL Buffered Substrate solution, 50mL Contrast BLUE solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.