Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RENBP (Human) ELISA Kit

Catalog No. 89100225 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Urine
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-225 CCS, Urine
89-028-594 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-225 Supplier Abnova™ Supplier No. KA0498
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human RENBP

Specifications

Calibration Range 0.031 to 2ng/mL
Conjugate Biotin
Cross Reactivity Human
Description RENBP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human RENBP.
Detection Limit 0.008 μg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Human RBP Standard (6μg), Biotinylated RBP Antibody (50x) (1 vial), EIA Diluent Concentrate (10x) (30mL), Wash Buffer Concentrate (20x) (30mL), Streptavidin-Peroxidase Conjugate (SP Conjugate) (90), Chromogen Substrate (8mL), Stop Solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type RENBP (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.