Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RETN (Human) ELISA Kit

Catalog No. 89100002 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
1 to 50ng/mL
78 to 5000pg/mL
Detection Limit:
0.012ng/mL
3pg/mL
Conjugate:
Biotin
HRP
Cross Reactivity:
Horse, Human, Monkey, Pig
Human
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range Detection Limit Conjugate Cross Reactivity
89-100-002 1 to 50ng/mL 0.012ng/mL HRP Horse, Human, Monkey, Pig
89-100-806 78 to 5000pg/mL 3pg/mL Biotin Human
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-002 Supplier Abnova™ Supplier No. KA0034
Add to Cart
Add to Cart

Includes

  • Antibody Coated Microtiter Strips, 96 wells
  • Biotin Labelled Antibody, 13mL
  • Streptavidin-HRP Conjugate, 13mL
  • Master Standard, 1 vial
  • Quality Control High, 1 vial
  • Quality Control Low, 1 vial
  • Dilution Buffer, 20mL
  • Wash Solution Conc. (10x), 100mL
  • Substrate Solution, 13mL
  • Stop Solution, 13mL

  • The kit measures total resistin in serum and plasma (EDTA, citrate, heparin)
  • Quality Controls are human serum based
  • Standard is recombinant protein based
  • Components of the kit are provided ready to use, concentrated or lyophilized

Specifications

Calibration Range 1 to 50ng/mL
Conjugate HRP
Cross Reactivity Horse, Human, Monkey, Pig
Description RETN (Human) ELISA Kit is a biotin labelled antibody based sandwich enzyme immunoassay for the quantitative measurement of human resistin in serum, plasma and tissue culture medium.
Detection Limit 0.012ng/mL
Detection Method Colorimetric
Includes Antibody Coated Microtiter Strips (96 wells), Biotin Labelled Antibody (13mL), Streptavidin-HRP Conjugate (13mL), Master Standard: 1 vial, Quality Control High: 1 vial, Quality Control Low: 1 vial, Dilution Buffer (20mL), Wash Solution Conc. (10x) (100mL), Substrate Solution (13mL), Stop Solution (13mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity 0.012 ng/mL
Total Assay Time <4 hr.
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.