Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ RETN (Human) ELISA Kit

Catalog No. 89100806 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
1 to 50ng/mL
78 to 5000pg/mL
Detection Limit:
0.012ng/mL
3pg/mL
Conjugate:
Biotin
HRP
Cross Reactivity:
Horse, Human, Monkey, Pig
Human
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range Detection Limit Conjugate Cross Reactivity
89-100-806 78 to 5000pg/mL 3pg/mL Biotin Human
89-100-002 1 to 50ng/mL 0.012ng/mL HRP Horse, Human, Monkey, Pig
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-806 Supplier Abnova™ Supplier No. KA3103
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Human Resistin antibody, 96 (8 x 12) wells
  • Lyophilized Resistin standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Human Resistin antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 78 to 5000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description RETN (Human) ELISA Kit is intended for the quantitative measurement of human RETN.
Detection Limit 3pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Human Resistin antibody 96 (8x12) wells, Lyophilized Resistin standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Human Resistin receptor antibody (Concentrated), Dilution: 1:100 (130mL), Antibody diluent buffer: 12mL, Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum, Tissue Lysates, Urine
Sample Volume 100 μL
Sensitivity <3 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type RETN (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.