Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Retn (Mouse) ELISA Kit

Catalog No. 89100807 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
156 to 10000pg/mL
31.2 to 2000pg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range
89-100-807 31.2 to 2000pg/mL
89-028-118 156 to 10000pg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-807 Supplier Abnova™ Supplier No. KA3104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Retn

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Retn (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Retn.
Detection Limit 2pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Mouse Resistin antibody, Lyophilized Resistin standards (100ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Mouse Resistin antibody (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated) (130uL), ABC diluent buffer (12mL),TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X)(30mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Retn (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.