Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rheumatoid Factor Ab ELISA Kit

Catalog No. 89028204 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-204 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-204 Supplier Abnova™ Supplier No. KA1442

Includes

  • Divisible microplate consisting of 12 modules of 8-wells each, coated Fc fragments of highly purified human Immunoglobulin G. Ready to use, Qty.1
  • Combined calibrators with IgG, IgM and IgA class rheumatoid factor antibodies (A-E) in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) containing: 0; 15; 50; 150; 500 U/mL. Ready to use, 5 vials, 1.5mL each
  • Rheumatoid factor controls in a serum/buffer matrix (PBS, BSA, NaN3 <0.1% (w/w)) positive (1) and negative (2), for the respective concentrations see the enclosed package insert. Ready to use, 2 vials, 1.5mL each
  • Sample buffer (Tris, NaN3 <0.1% (w/w)), yellow, concentrate (5x), 1 vial, 20mL
  • Enzyme conjugate solution (PBS, Proclin 300 <0.5% (v/v)), (light red) containing polyclonal rabbit anti-human IgG, polyclonal rabbit anti-human IgM and polyclonal rabbit anti-human IgA; labelled with horseradish peroxidase, Ready to use, 1 vial, 15mL

Indirect Solid Phase ELISA for the quantitative measurement of IgG, IgM and IgA class Rheumatoid factor antibodies

Specifications

Conjugate HRP
Cross Reactivity Human
Description Rheumatoid Factor Ab (Human) ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG, IgM and IgA class rheumatoid factor antibodies.
Detection Limit 1U/mL
Detection Method Colorimetric
Includes 96-well plate, 5 vials, 1.5mL each Combined calibrator with IgG, IgM and IgA class rheumatoid factor Antibodies, 2 vials, 1.5mL each Rheumatoid factor controls in a Serum/Buffer Matrix, 1 vial, 20mL Sample Buffer, 1 vial, 15mL Enzyme Conjugate solution, 1 vial, 15mL TMB Substrate solution, 1 vial, 15mL stop solution, 1 vial, 20mL wash solution
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage 4°C
For Use With (Application) Quantification
Product Type Rheumatoid Factor Ab ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.