Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rheumatoid factor IgA ELISA Kit

Catalog No. 89024581 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-581 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-581 Supplier Abnova™ Supplier No. KA1289
Add to Cart
Add to Cart

Indirect Solid Phase Enzyme Immunoassay for the quantitative measurement of Rheumatoid factor IgG in Serum and Plasma

Specifications

Conjugate HRP
Cross Reactivity Human
Description Rheumatoid factor IgA ELISA Kit is indirect solid phase enzyme immunoassay for the quantitative measurement of rheumatoid factor IgA in serum and plasma.
Detection Method Colorimetric
Includes 12 Goat IgG coated microtiter strip, 4 X 2mL standard with 0, 50, 200 and 500 IU/mL, 2mL positive Control, 2mL negative Control, 15mL Enzyme Conjugate, 15mL Substrate, 15mL stop solution, 60mL sample diluent, 60mL (10X) Washing Buffer
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Rheumatoid factor IgA ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.