Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ S100B (Human) ELISA Kit

Catalog No. 89022532 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-532 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-532 Supplier Abnova™ Supplier No. KA0037
Add to Cart
Add to Cart

A HRP labelled antibody based sandwich enzyme immunoassay for the quantitative measurement of human S100B protein only for serum samples, heparin plasma samples and cerebrospinal fluid samples

  • The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs
  • S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation
  • S100 genes include at least 13 members which are located as a cluster on chromosome 1q21; however, this gene is located at 21q22.3
  • This protein may function in Neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation and inhibition of microtubule assembly
  • Chromosomal rearrangements and altered expression of this gene have been implicated in several neurological, neoplastic and other types of diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma and type I diabetes

Specifications

Antigen S100B
Calibration Range 0.05 to 2ng/mL
Conjugate HRP
Cross Reactivity Bovine, Cat, Goat, Hamster, Human, Rabbit, Rat
Description S100B (Human) ELISA Kit is a HRP labelled antibody based sandwich enzyme immunoassay for the quantitative measurement of human S100B protein just only for serum samples, heparin plasma samples and cerebrospinal fluid samples.
Detection Method Colorimetric
Includes 96-well Antibody coated microtiter strip, 0.15mL (100X) Conc. Biotin Labelled Antibody, 13mL Streptavidin HRP Conjugate, Master standard 1 vial, Quality control high 1 vial, Quality control low 1 vial, 13mL Biotin-Ab Diluent, 20mL Dilution Buffer, 100mL conc. (10X) Wash solution, 13mL Substrate solution, 13mL stop solution
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Cerebrospinal Fluid, Plasma (Heparin), Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type S100B (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.