Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SCARB2 (Human) ELISA Kit

Catalog No. 89100458 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-458 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-458 Supplier Abnova™ Supplier No. KA2178

Includes

  • LIMP-II Microplate: 96 wells coated with anti-human LIMP-II
  • Wash Buffer Concentrate (20x)
  • Standards: recombinant human LIMP-II
  • Assay Diluent C: 30mL diluent buffer. For Standard/Sample (serum/plasma) diluent
  • Assay Diluent B: 15mL of 5x concentrated buffer. For Standard/Sample (cell culture medium/urine) diluent
  • Detection Antibody: 2 vial of biotinylated anti-human LIMP-II
  • HRP-Streptavidin Concentrate: 8μL 10,000x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent: 12mL of 3,3',5,5'-tetramethylbenzidine (TMB) in buffer solution
  • Stop Solution: 2M sulfuric acid

Use to measure quantitatively human SCARB2. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for human LIMP-II coated on a 96-well plate

Specifications

Conjugate HRP
Cross Reactivity Human
Description SCARB2 (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of human SCARB2.
Detection Limit 10pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma, Cell Culture Supernatant
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type SCARB2 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.