Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SCGB1A1 (Human) ELISA Kit

Catalog No. 89100010 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Plasma (Citrate, EDTA, Heparin), Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-010 Plasma (Citrate, EDTA, Heparin), Serum
89-101-041 CCS, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-010 Supplier Abnova™ Supplier No. KA0020
Add to Cart
Add to Cart

Includes

  • Antibody Coated Microtiter Strips, 96 wells
  • Biotin Labelled Antibody, 13mL
  • Streptavidin-HRP Conjugate, 13mL
  • Master Standard, 2 vials
  • Quality Control HIGH, 2 vials
  • Quality Control LOW, 2 vials
  • Dilution Buffer, 20mL
  • Wash Solution Conc. (10x), 100mL
  • Substrate Solution, 13mL
  • Stop Solution, 13mL

  • The kit measures Clara cell protein in serum and plasma (EDTA, citrate, heparin)
  • Quality Controls are human serum based
  • Standard is recombinant protein (E. coli) based
  • Components of the kit are provided ready to use, concentrated or lyophilized

Specifications

Calibration Range 1.57 to 50ng/mL
Conjugate HRP
Cross Reactivity Human
Description SCGB1A1 (Human) ELISA Kit is a biotin labelled antibody based sandwich enzyme immunoassay for the quantitative measurement of human club cell protein.
Detection Limit 46pg/mL
Detection Method Colorimetric
Includes Antibody Coated Microtiter Strips 96 wells, Biotin Labelled Antibody (13mL), Streptavidin-HRP Conjugate (13mL), Master Standard (1 vial), Quality Control HIGH (2 vials), Quality Control LOW (2 vials), Dilution Buffer (20mL), Wash Solution Conc. (10x) (100mL), Substrate Solution (13mL), Stop Solution (13mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity 46 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.