Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SCGB1A1 (Human) ELISA Kit

Catalog No. 89101041 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Plasma (Citrate, EDTA, Heparin), Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-101-041 CCS, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
89-100-010 Plasma (Citrate, EDTA, Heparin), Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-041 Supplier Abnova™ Supplier No. KA3362
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-human SCGB1A1 antibody, 96 (8 x 12) wells
  • Lyophilized recombinant human SCGB1A1 standard, 10ng x 2
  • Biotinylated anti-human SCGB1A1 antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

SCGB1A1 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description SCGB1A1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SCGB1A1.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well plate precoated with anti- human SCGB1A1 antibody 96 (8x12) wells, Lyophilized recombinant human SCGB1A1 standard (10ng x 2), Biotinylated anti- human SCGB1A1antibody(dilution 1:100) (130μL), Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100) (130μL), Sample diluent buffer (30mL), Antibody diluent buffer (12mL), BC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Human Milk, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity Natural and recombinant human SCGB1A1
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.