Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Secreted Alkaline Phosphatase Reporter Gene Assay Kit (Luminescence)

Catalog No. 89024643 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-643 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-643 Supplier Abnova™ Supplier No. KA1362
Add to Cart
Add to Cart

Used for the sensitive quantitation of SEAP in conditioned cell culture medium from transfected cells and functional study

Specifications

Product Type Reporter Gene Assay Kit
Description Secreted Alkaline Phosphatase Reporter Gene Assay Kit (Luminescence) provides a simple chemiluminescence method for the sensitive quantitation of SEAP in conditioned cell culture medium from transfected cells.
Includes 15mL SEAP substrate (Luminescence), 50μL Alkaline Phosphatase standard, 3 plates 96-well solid plate (white) with lid
Content And Storage Store the kit at 4°C temporarily. For best results, store the components as described in the protocol.
For Use With (Application) Functional Study
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.