Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Serotonin ELISA Kit

Catalog No. 89028627 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Biological sample
Platelet, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-028-627 Platelet, Serum, Urine
89-100-615 Biological sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-627 Supplier Abnova™ Supplier No. KA1894
Add to Cart
Add to Cart

Enzyme immunoassay for the quantitative determination of Serotonin in serum, urine and platelets

  • Sample Type: Platelet, Serum, Urine
  • Sample Volume: 25μL

Applications:

Quantification

Specifications

Calibration Range 40 to 860ng/mL
Description Serotonin ELISA Kit is an enzyme immunoassay for the quantitative determination of Serotonin in serum, urine and platelets.
Detection Limit 5ng/mL
Detection Method Colorimetric
Includes 2 x 50 Reaction tubes, 1 x 20mL Wash Buffer concentrate, 1 x 12mL Conjugate Anti-Rabbit IgG Conjugated with peroxidase, 1 x 12mL Substrate, containing a solution of Tetramethylbenzidine (TMB), 1 x 12mL stop solution, containing 0.25 M H2SO4, 1 x 96-well Serotonin-5-HIAA microtiter strip 12 strip, 8-well each, break apart, Precoated, 1 x 4mL standard A, 1 x 12mL, 1 x 4mL standard B, 1 x 4mL standard C, 1 x 4mL standard D, 1 x 4mL standard E, 1 x 4mL standard F, 1 x 12mL Serotonin Antiserum from Rabbit, 1 x 55mL Acylation Buffer, 1 x 3mL Acylation Reagent, 1 x 4mL Control 1, 1 x 4mL Control 2
Quantity 1 Kit
Sample Type Platelet, Serum, Urine
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Serotonin ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.