Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SERPINA1 (Human) ELISA Kit

Catalog No. 89100251 Shop All Abnova Corporation Products
Change view
Click to view available options
Calibration Range:
0.039 to 40 μg/mL
0.391 to 100 ng/mL
Detection Limit:
0.018 μg/mL
0.16 ng/mL
Sample Volume:
25 μL
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Range Detection Limit Sample Volume
89-100-251 0.391 to 100 ng/mL 0.16 ng/mL 50 μL
89-100-252 0.039 to 40 μg/mL 0.018 μg/mL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-251 Supplier Abnova™ Supplier No. KA0459
Add to Cart
Add to Cart

Includes

  • Human A1AT Microplate: A polystyrene microplate coated with a polyclonal antibody against human A1AT
  • Sealing Tapes
  • Human A1AT Standard: Human A1AT in a buffered protein base, lyophilized, 150ng
  • Biotinylated Human A1AT Antibody (50x): A 50-fold biotinylated polyclonal antibody against human A1AT, 120μL
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate, 80μL
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

This assay employs a quantitative sandwich enzyme immunoassay technique that measures A1AT in less than 4 hours

Specifications

Calibration Range 0.391 to 100 ng/mL
Conjugate Biotin
Cross Reactivity Human
Description SERPINA1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SERPINA1 in cell culture supernatant.
Detection Limit 0.16 ng/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Human A1AT Standard (400ng), Biotinylated A1AT Antibody (100x) (1 vial), MIX Diluent Concentrate (10x) (30mL), Wash Buffer Concentrate (20x) (30mL x 2), Streptavidin-Peroxidase Conjugate (SP Conjugate) 80, Chromogen Substrate (8mL), Stop Solution: (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Samples, CSF, Milk, Saliva, Urine
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.