Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SERPINC1 (Human) ELISA Kit

Catalog No. 89100254 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Urine
Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-254 CCS, Urine
89-028-588 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-254 Supplier Abnova™ Supplier No. KA0456
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human SERPINC1

Specifications

Calibration Range 3.125 to 200ng/mL
Conjugate Biotin
Cross Reactivity Human
Description SERPINC1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SERPINC1.
Detection Limit 2ng/mL
Detection Method Colorimetric
Includes 96 wells-Antibody coated microwells, AT III Standard (400ng), 100X Biotinylated AT III Antibody (1vial), 10X MIX Diluent Concentrate (30mL), 20X Wash Buffer Concentrate (2 x 30mL), 80 Streptavidin-Peroxidase Conjugate (SP Conjugate), Chromogen substare (8mL), Stop solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Urine
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type SERPINC1 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.