Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SFTPD (Human) ELISA Kit

Catalog No. 89025090 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-025-090 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-025-090 Supplier Abnova™ Supplier No. KA1251
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human SFTPD in Serum, Plasma (-EDTA, -Heparin, -Citrate), Amniotic fluid, bronchoalveolar lavage

Specifications

Calibration Range 1.56 to 100ng/mL
Conjugate HRP
Cross Reactivity Bovine, Horse, Human, Monkey
Description SFTPD (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SFTPD in serum, plasma (-EDTA, -Heparin, -Citrate), amniotic fluid, bronchoalveolar lavage.
Detection Limit 0.01ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microtiter strip, 13mL Biotin labelled Antibody,13mL Streptavidin HRP Conjugate, 1 vial Master standards, 2 vials Quality control high, 2 vials Quality control low, 50mL Dilution Buffer, 100mL Conc. (10X) Wash solution, 13mL Substrate solution, 13mL stop solution
Quantity 1 Kit
Sample Type Amniotic Fluid, Bronchoalveolar Lavage fluid, Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type SFTPD (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.