Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SHBG (Human) ELISA Kit

Catalog No. 89028725 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-725 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-725 Supplier Abnova™ Supplier No. KA1913
Add to Cart
Add to Cart

Enzyme Immunoassay for the quantitative measurement of SHBG in Human Serum

Specifications

Conjugate HRP
Cross Reactivity Human
Description SHBG (Human) ELISA Kit is an enzyme immunoassay for the quantitative measurement of SHBG in human serum.
Detection Limit 0.1nM/L
Detection Method Colorimetric
Includes 1 x 96-well microtiter strip, 1 x 50mL Wash Buffer concentrate, 1 x 0.4mL HRP Conjugate, 6 vials SHBG calibrator, 0.4mL Control, 1x 40mL Assay Buffer, 1 x 16mL TMB Substrate, 1 x 16mL Stopping solution
Quantity 1 Kit
Sample Type Serum
Sample Volume 20 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type SHBG (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.