Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SHH (N-terminal) (Human) ELISA Kit

Catalog No. 89100424 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-424 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-424 Supplier Abnova™ Supplier No. KA2144

missing translation for 'validMainframeMsg'

Includes

  • Shh-N Microplate (Item A): 96 wells coated with anti-human Shh-N
  • Wash Buffer Concentrate (20x) (Item B)
  • Standards (Item C): Recombinant human Shh-N
  • Assay Diluent C (Item L): Diluent buffer, 0.09% sodium azide as preservative. For Standard/Sample (serum/plasma) diluent
  • Assay Diluent B (Item E): 5x concentrated buffer. For Standard/Sample (cell culture medium/urine) diluent
  • Detection Antibody (Item F): Biotinylated anti-human Shh-N
  • HRP-Streptavidin Concentrate (Item G): 200x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H)
  • Stop Solution (Item I): 0.2M sulfuric acid

Use to measure quantitatively human SHH N-terminal. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for human Shh-N coated on a 96-well plate

Specifications

Conjugate HRP
Cross Reactivity Human
Description SHH (N-terminal) (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of human SHH N-terminal.
Detection Limit 42pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma, Cell Culture Supernatant, Urine
Sample Volume 100 μL
Sensitivity The minimum detectable dose of Shh to N is typically less than 42 pg/mL in Assay Diluent C and 8 pg/mL in Assay Diluent B respectivelly
Packaging 96-well
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type SHH (N-terminal) (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.