Learn More
Description
Includes
- Assay plate, 1
- Standard, 2
- Sample diluent, 1 x 20mL
- Assay diluent A, 1 x 10mL
- Assay diluent B, 1 x 10mL
- Detection reagent A, 1 x 120μL
- Detection reagent B, 1 x 120μL
- Wash buffer (25 x concentrate), 1 x 30mL
- Substrate, 1 x 10mL
- Stop solution, 1 x 10mL
- Plate sealer for 96 wells, 5
SPARC (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SPARC.
Quantification
Specifications
Specifications
| Calibration Range | 3.12 to 200ng/mL |
| Conjugate | HRP |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Includes | 1 Assay plate, 20mL standard, 20mL sample diluent, 10mL assay diluent A, 10mL assay diluent B, 120μL detection reagent A, 120μL detection reagent B, 30mL 25X Wash Buffer, 10mL Substrate, 10mL stop solution, 5 plate seal |
| Quantity | 96 Assays |
| Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Homogenate |
| Sample Volume | 100 μL |
| Content And Storage | 4°C |
| For Use With (Application) | For the quantitative measurement of Human SPARC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.