Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Spermatozoa Ab ELISA Kit

Catalog No. 89025095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-025-095 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-025-095 Supplier Abnova™ Supplier No. KA1256
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative determination of Anti-spermatozoa antibodies in Human Serum

Specifications

Calibration Range 0 to 60U/mL
Conjugate HRP
Cross Reactivity Human
Description Anti-Spermatozoa Ab ELISA Kit is a solid-phase sandwich enzyme-immunoassay for the quantitative determination of anti-spermatozoa antibodies in human serum.
Detection Method Colorimetric
Includes 4 Vial, 0.5mL Sperm Antibody ELISA standard set, 1 vial, 0.5mL Control, 1 vials, 50mL Dilution Buffer, 1 vials, 50mL Washing solution, 1 vial, 8mL Enzyme Conjugate, 1 vial, 13mL Substrate solution, 1 vial, 13mL stop solution, 1 plate 96-well plate, 1 x holder for single strip
Quantity 1 Kit
Sample Type Serum
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Spermatozoa Ab ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.