Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SPP1 (Human) ELISA Kit

Catalog No. 89100282 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-100-282 Biotin
89-100-927 HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-282 Supplier Abnova™ Supplier No. KA0407
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-human OPN antibody, 96 (8 x 12) wells
  • Lyophilized recombinant human OPN standard, 10ng/tube x 2
  • Biotinylated anti-human OPN antibody, dilution 1:100, 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

The SPP1 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 156 to 10000ng/mL
Conjugate Biotin
Cross Reactivity Human
Description SPP1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SPP1.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Lyophilized recombinant Human SPP1 standard (10ng x 2 vials), Sample diluent buffer (30mL), Biotinylated anti-Human SPP1 antibody (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase complex (ABC): 130μL, ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Milk, Plasma (EDTA, Heparin), Serum, Urine
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity Nature and recombinant human OPN
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.