Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SPP1 (Human) ELISA Kit

Catalog No. 89100927 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-100-927 HRP
89-100-282 Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-927 Supplier Abnova™ Supplier No. KA3390
Add to Cart
Add to Cart

Includes

  • Clear Coated 96 Well Plate: Clear plastic microplate with break-apart strips coated with mouse anti-human Osteopontin, 1 plate
  • Osteopontin Standard: recombinant human Osteopontin at 20ng, 2 vials
  • Osteopontin Conjugate: A monoclonal antibody to human Osteopontin labeled with peroxidase, 5mL
  • Assay Buffer Concentrate: A 5X concentrate, 28mL
  • Wash Buffer Concentrate: A 20X concentrate, 30mL
  • TMB Substrate, 11mL
  • Stop Solution: A 1N hydrochloric acid solution. Caustic, 5mL
  • Plate Sealer, 1 unit

The SPP1 (Human) ELISA Kit is designed to quantitatively measure human Osteopontin present in biological samples and tissue culture media

Specifications

Calibration Range 0 to 40ng/mL
Conjugate HRP
Cross Reactivity Human
Description SPP1 (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of SPP1 in biological samples and tissue culture media.
Detection Limit 0.248ng/mL
Detection Method Colorimetric
Includes Clear Coated 96 Well Plate: Clear plastic microplate with break-apart strips coated with mouse anti-human Osteopontin. (1plate), Osteopontin Standard: recombinant human Osteopontin at 20 ng. (2vials), Osteopontin Conjugate: A monoclonal antibody to human Osteopontin labeled with peroxidase (5mL), Assay Buffer Concentrate: A 5X concentrate that should be diluted with deionized or distilled water (28mL), Wash Buffer Concentrate: A 20X concentrate that should be diluted with deionized or distilled water (30mL), TMB Substrate (11mL), Stop Solution: A 1N hydrochloric acid solution. Caustic. (5mL), Plate Sealer (1unit)
Regulatory Status For research use only (RUO)
Sample Type Milk, Plasma (EDTA), Tissue Culture Medium, Urine
Sample Volume 50 μL
Sensitivity 0.246 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type SPP1 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.