Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SRBC IgG (Rat) ELISA Kit

Catalog No. 89100684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-684 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-684 Supplier Abnova™ Supplier No. KA2494
Add to Cart
Add to Cart

Includes

  • SRBC coated 96-well plate, 96 wells (12 strips of 8 wells)
  • Reference Standard stock, Lyophilized
  • Diluent, 30mL
  • Enzyme Conjugate Reagent, 11mL
  • 20x Wash Solution, 50mL
  • TMB Reagent (One-step), 11mL
  • Stop Solution (1N HCl), 11mL

Use to measure quantitatively SRBC IgG in plasma and serum.

SRBC IgG (Rat) ELISA Kit is based on a solid phase enzyme-linked immunosorbent assay (ELISA)

Specifications

Calibration Range 6.25 to 100U/mL
Conjugate HRP
Cross Reactivity Rat
Description SRBC IgG (Rat) ELISA Kit is for the quantitative measurement of SRBC IgG in plasma and serum.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type SRBC IgG (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.