Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SREBF1 Transcription Factor Assay Kit

Catalog No. 89024651 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-651 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-651 Supplier Abnova™ Supplier No. KA1377
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SREBF1 Transcription Factor Assay Kit is a method for detecting SREBF1 transcription factor DNA binding activity in nuclear extracts.

  • A 96-well enzyme-linked immunosorbent assay (ELISA) replaces the cumbersome radioactive electrophoretic mobility shift assay (EMSA)
  • A specific double stranded DNA (dsDNA) sequence containing the SREBP response element is immobilized onto the wells of a 96-well plate
  • SREBP-1 contained in a nuclear extract binds specifically to the SREBP response element
  • SREBF1 Transcription Factor Assay Kit detects human, rat, and murine SREBP-1
  • Kit components may be stored at -20°C prior to use
  • This product is for laboratory research use only: not for administration to humans

Specifications

Description SREBF1 Transcription Factor Assay Kit is a method for detecting SREBF1 transcription factor DNA binding activity in nuclear extracts.
Immunoassay Kit Format ELISA (Standard)
Product Type SREBF1 Transcription Factor Assay Kit
Sufficient For 15 Reactions
For Use With (Application) Functional Study
Gene ID (Entrez) 6720
Gene Symbol SREBF1
Quantity 1 Kit
Storage Requirements Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.