Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SS-B (La) (Human) IgG ELISA Kit

Catalog No. 89024491 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-491 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-491 Supplier Abnova™ Supplier No. KA0950
Add to Cart
Add to Cart

For the detection of IgG antibody to SSB in Human Serum or Plasma

Specifications

Conjugate HRP
Cross Reactivity Human
Description SSB (La) (Human) IgG ELISA Kit is used for the detection of IgG antibody to SSB in human serum or plasma.
Detection Method Colorimetric
Includes Microwell 12 x 8 x 1, 22mL sample diluent, 1.5mL Cut-off calibrator, 1.5mL positive Control, 1.5mL negative Control, 12mL Enzyme Conjugate, 12mL TMB Chromogenic Substrate, 12mL stop solution, 25mL (20X) Washing concentrate
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 10 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Qualitative
Product Type SS-B (La) (Human) IgG ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.