Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ STAT1 (pS727) (Human/Mouse) ELISA Kit

Catalog No. 89100451 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-451 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-451 Supplier Abnova™ Supplier No. KA2171

Includes

  • Stat1 (Ser727) Microplate (Item A): 96 wells coated with anti-Stat1 (Ser727)
  • Wash Buffer Concentrate (20x) (Item B)
  • Assay Diluent (Item E)
  • Detection Antibody Stat1 (Item C): 2 vial of biotinylated anti-Stat1
  • HRP-Streptavidin Concentrate (Item G): 8μL concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H)
  • Stop Solution (Item I): 2M sulfuric acid
  • Cell Lysate Buffer (Item J): 5mL 2x cell lysis buffer (not including protease and phosphatase inhibitors)
  • Positive Control A431S001-2 (Item K): lyophilized powder from A431 cell lysate

Use to detect qualitatively human and mouse phospho STAT1 (S727). Enzyme-linked immunosorbent assay.

STAT1 (pS727) (Human/Mouse) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates

Specifications

Conjugate HRP
Cross Reactivity Human, Mouse
Description STAT1 (pS727) (Human/Mouse) ELISA Kit is an enzyme-linked immunosorbent assay for the qualitative determination of human and mouse phospho STAT1 (S727).
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Lysate, Tissue Lysate
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Qualitative
Product Type STAT1 (pS727) (Human/Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.