Learn More
Description
Includes
- Stat1 Microplate (Item A): Coated with anti-phospho-Stat1 (Ser727) (half plate) and anti-Stat1 antibody (half plate)
- Wash Buffer Concentrate (20x) (Item B)
- Assay Diluent (5x) (Item E)
- Detection Antibody Stat1 (Item C): Biotinylated anti-Stat1
- HRP-Streptavidin Concentrate (Item G): HRP-conjugated streptavidin concentrate
- TMB One-Step Substrate Reagent (Item H)
- Stop Solution (Item I): 0.2M sulfuric acid
- Cell Lysate Buffer (Item J): 2x cell lysis buffer (not including protease and phosphatase inhibitors)
- Positive Control A431S002-1 (Item K): Lyophilized powder from A431 cell lysate
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human, Mouse |
| Description | STAT1 (pS727)/Pan STAT1 (Human/Mouse) ELISA Kit is an enzyme-linked immunosorbent assay for the qualitative determination of human, mouse phospho STAT1 (S727) and Pan STAT1. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Regulatory Status | For research use only (RUO) |
| Sample Type | Cell Lysate, Tissue Lysate |
| Sample Volume | 100 μL |
| Packaging | 96-well |
| Content And Storage | Store the kit at -20°C. For best results, store the components as described in the protocol. Avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.