Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Swine H1N1 HA ELISA Kit

Catalog No. 89100361 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-361 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-361 Supplier Abnova™ Supplier No. KA1292
Add to Cart
Add to Cart

Includes

  • Swine H1N1 Antibody Coated Stripwell Microplate, 96-wells (8-well Strips x 12)
  • Detection Antibody, 10μL/vial x 1
  • Streptavidin-HRP, 5μL/vial x 1
  • Protein Standard Stock, 2μg/vial x 1
  • TMB Substrate Solution, 11mL/bottle x 1
  • Diluent, 40mL/bottle x1
  • 10x Solution A, 30mL/bottle x 1
  • 10x Solution B, 50mL/bottle x 1
  • Adhesive plastic, 3Pcs.
  • Stop Solution, 10mL/bottle x 1

Use to measure quantitatively H1N1 HA. Enzyme-linked immunosorbent assay.

The design of this assay is based on a sandwich Enzyme-Linked Immunosorbent Assay (ELISA) for the quantitative determination of swine H1N1 (A/California/04/2009) HA

Specifications

Calibration Range 0.12 to 10ng/mL
Conjugate Biotin
Description Swine H1N1 HA ELISA Kit is an Enzyme-Linked Immunosorbent Assay for the quantitative measurement of H1N1 HA.
Detection Limit 0.12ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sensitivity 0.12 ng/mL
Packaging 96-well
Content And Storage Store the Detection Antibody and Protein Standard Stock at -80°C, and aliquot to minimize the repeat freeze-thaw cycle. Store other components at 4°C. TMB should be keep away from light.
For Use With (Application) Quantification
Product Type Swine H1N1 HA ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.