Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ T3 (Free) (Human) ELISA Kit

Catalog No. 89100042 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-042 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-042 Supplier Abnova™ Supplier No. KA0199
Add to Cart
Add to Cart

Includes

  • Antibody-Coated Microplate: Microtiter wells coated with Anti-T3
  • fT3-Enzyme Conjugate Reagent, ready to use: Contain T3 Ab conjugated to horseradish perosidase with preservatives
  • Free T3 Reference Standards Set: Contains 0, 1.6, 3.6, 6.0, 9.2 and 20pg/mL of fT3 in human serum with preservatives, liquid, ready to use, Note: Exact levels are given on the labels on a lot specific basis, 1mL x 6
  • Color Reagent A; Contains hydrogen peroxide in acetate buffer, 13mL
  • Color Reagent B Contains 3,3',5,5' tetramethylbenzidine (TMB) stabilized in buffer solution, 13mL
  • Stop Solution (3N HCl); Contains diluted hydrochloric acid

Use to measure quantitatively free triiodothyronine. Sandwich enzyme immunoassay.

The fT3 test is a solid phase competitive enzyme immunoassay

Specifications

Calibration Range 0 to 20pg/mL
Conjugate HRP
Cross Reactivity Human
Description T3 (Free) (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of free triiodothyronine.
Detection Limit 0.05pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 50 μL
Sensitivity 0.05 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.