Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ T3 (Total) (Human) ELISA Kit

Catalog No. 89022679 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-679 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-679 Supplier Abnova™ Supplier No. KA0198
Add to Cart
Add to Cart

Includes

  • Goat Anti-Mouse IgG Coated Microtiter Wells, 96 wells, Ready to use, 1 in number
  • Enzyme Conjugate Concentrate (11x), Concentrated, 1.3mL
  • Enzyme Conjugate Diluent, Ready to use, 13mL
  • T3 Reference Standard, 0, 0.75, 1.5, 3.0, 6.0 and 10.0ng/mL, 1 set, 1.0 mL each, ready to use, 1mL each
  • Antibody Reagent, ready to use, 7mL
  • TMB Reagent (One-Step), Ready to use, 11mL
  • Stop Solution (1N HCl), Ready to use, 11mL

Sandwich Enzyme Immunoassay for the quantitative measurement of total Triiodothyronine

Specifications

Calibration Range 0 to 10ng/mL
Conjugate HRP
Cross Reactivity Human
Description T3 (Total) (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of total triiodothyronine.
Detection Method Colorimetric
Includes Goat Anti-Mouse IgG coated microtiterwell, 96-well 1, 1.3mL (11x) Enzyme Conjugate concentrate, 13mL Enzyme Conjugate Diluent, T3 Reference standard, 0, 0.75, 1.5, 3, 6 and 10ng/mL, 1 set, 1mL each, ready to use, 7mL Antibody reagent, 11mL TMB reagent (one-step), 11mL stop solution (1NHCl)
Quantity 1 Kit
Sample Type Plasma (EDTA, Heparin, Oxalte)
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type T3 (Total) (Human) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.