Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ T3 (Total) (Mouse/Rat) ELISA Kit

Catalog No. 89024466 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-466 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-466 Supplier Abnova™ Supplier No. KA0925
Add to Cart
Add to Cart

For the quantitative measurement of Triiodothyronine (T3) in Mouse/Rat Serum or Plasma

Specifications

Calibration Range 0.25 to 7.5ng/mL
Conjugate HRP
Cross Reactivity Mouse, Rat
Description T3 (Total) (Mouse/Rat) ELISA Kit is used for the quantitative measurement of triiodothyronine (T3) in mouse/rat serum or plasma.
Detection Limit 0.186ng/mL
Detection Method Colorimetric
Includes Microwell coated with T3 polyclonal Ab 96 (12 x 8) well, 0.25mL T3 standard 7 vials, 1.5mL T3 Enzyme Conjugate concentrate 1 vial, 12mL Assay diluent, 12mL TMB Substrate 1 Bottle, 12mL stop solution 1 Bottle, 25mL Wash concentrate 1 Bottle (20X)
Quantity 96 Assays
Sample Type Plasma, Serum
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type T3 (Total) (Mouse/Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.