Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ T4 (Rodent) ELISA Kit

Catalog No. 89100594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-594 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-594 Supplier Abnova™ Supplier No. KA2335
Add to Cart
Add to Cart

Includes

  • Antibody-coated microtiter wells, 96 wells
  • Lyophilized Standard (0, 5, 10, 25, 50, 100, 300ng/mL) reconstitute in 1mL standard/sample diluent, 1 set (7 vials)
  • HRP Conjugate Reagent, 12mL
  • TMB Color Reagent, ready for use, 12mL
  • Stop solution (2N HCl), 6mL
  • 20x Wash buffer, 20mL
  • Standard/Sample diluent, 20mL

Use to measure quantitatively thyroxin (T4) in rodent serum, plasma. Immunoassay.

In the T4 ELISA Test, a certain amount of anti-T4 antibody is coated on microtiter wells

Specifications

Calibration Range 0 to 300ng/mL
Conjugate HRP
Cross Reactivity Rodents
Description T4 (Rodent) ELISA Kit is an immunoassay designed for the quantitative determination of thyroxin (T4) in rodent serum, plasma.
Detection Limit 2ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 50 μL
Sensitivity 2 ng/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.