Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ T4 (Total) (Human), HRP, ELISA Kit

Catalog No. 89100041 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-041 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-041 Supplier Abnova™ Supplier No. KA0200
Add to Cart
Add to Cart

Includes

  • Sheep anti-T4 coated microtiter wells 96 wells
  • T4 Reference Standards: 0, 2, 5, 10, 15, and 25 ug/dl, ready to use 1 set, 1ml/vial
  • Enzyme Conjugate Reagent Concentrate (11x) 1.3mL
  • Enzyme Conjugate Diluent 13mL
  • TMB Reagent (One-Step) 11mL
  • Stop Solution (1N HCl) 11mL

For quantitative measurement of Mouse Adiponectin/Acrp30 protein in serum and tissue culture medium

Specifications

Calibration Range 0 to 25μg/dL
Conjugate HRP
Cross Reactivity Human
Description T4 (Total) (Human) ELISA Kit is a competitive enzyme immunoassay for the quantitative measurement of total thyroxine.
Detection Limit 0.4 μg/dL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 25 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type T4 (Total) (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.