Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Testosterone (Horse) ELISA Kit

Catalog No. 89100565 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-565 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-565 Supplier Abnova™ Supplier No. KA2306
Add to Cart
Add to Cart

Includes

  • Antibody-coated microtiter wells, ready for use, 96 wells/plate
  • Reference Standard (0, 0.1, 0.5, 1.0, 2.0, 10, 20ng/mL) QC1 (∼0.5ng/mL) and QC2 (∼ 2.0ng/mL), ready for use, 0.5mL/vial
  • Enzyme Conjugate Reagent, ready for use, 12mL
  • TMB Color Reagent, ready for use, 12mL
  • Stop solution (2N HCl), ready for use, 6mL
  • 20x Wash buffer, ready for use, 20mL

Use to measure quantitatively testosterone in horse serum, plasma. Enzyme immunoassay.

The Testosterone quantitative test is based on a solid-phase enzyme immunoassay based on competitive binding method

Specifications

Calibration Range 0 to 20ng/mL
Conjugate HRP
Cross Reactivity Horse
Description Testosterone (Horse) ELISA Kit is an enzyme immunoassay for quantitative determination of testosterone in horse serum, plasma.
Detection Limit 20pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma
Sample Volume 50 μL
Sensitivity 20 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.