Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Testosterone (Pig) ELISA Kit

Catalog No. 89100608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-608 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-608 Supplier Abnova™ Supplier No. KA2349
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Microtiter wells coated with anti-testosterone antibody, 96 wells/plate
  • Enzyme conjugate reagent, 12mL
  • Testosterone reference standard set: 0, 0.1, 0.5, 1.0, 2.0, 10, 20ng/mL, lyophilized, should be diluted in 1mL standard/sample diluent, 1 vial x 7
  • QC1 (∼ 0.5ng/mL) should be diluted in 1mL standard/sample diluent, 1 vial
  • QC2 (∼ 2.0ng/mL) should be diluted in 1mL standard/sample diluent, 1 vial
  • TMB Color Reagent, 12mL
  • Stopping solution, 6mL
  • 20x Wash buffer, 20mL
  • Standard/sample diluent, 20mL

Use to measure quantitatively testosterone in Pig serum, plasma. Enzyme Immunoassay.

The Testosterone (Pig) ELISA Kit is based on a solid-phase enzyme immunoassay based on competitive binding method

Specifications

Calibration Range 0.1 to 20ng/mL
Conjugate HRP
Cross Reactivity Pig
Description Testosterone (Pig) ELISA Kit is an enzyme immunoassay for quantitative determination of testosterone in Pig serum, plasma.
Detection Limit 20pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum, Urine
Sample Volume 50 μL
Sensitivity 20 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C. Do not freeze.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.