Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Testosterone ELISA Kit

Catalog No. 89100933 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-933 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-933 Supplier Abnova™ Supplier No. KA3396
Only null left
Add to Cart
Add to Cart

Includes

  • Coated Clear 96 Well Plates: Clear plastic microtiter plate coated with goat anti-rabbit IgG, 1 plate
  • Testosterone Standard: Testosterone at 200,000pg/mL in a special stabilizing solution, 70μL
  • Testosterone Antibody: A rabbit polyclonal antibody specific for testosterone, 3mL
  • Testosterone Conjugate: A testosterone-peroxidase conjugate in a special stabilizing solution, 3mL
  • Assay Buffer Concentrate: A 5X concentrate, 28mL
  • Wash Buffer Concentrate: A 20X concentrate
  • TMB Substrate, 11mL
  • Stop Solution: A 1M solution of hydrochloric acid. CAUSTIC, 5mL
  • Plate Sealer, 1 slice

Use to detect quantitatively of testosterone in in urine and fecal samples. Enzyme-linked immunosorbent assay.

Testosterone ELISA Kit uses a specifically generated antibody to measure testosterone and its metabolites in urine and fecal samples, or in extracted serum and plasma

Specifications

Calibration Range 0 to 10,000pg/mL
Conjugate HRP
Description Testosterone ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of testosterone in in urine and fecal samples.
Detection Limit 30.6pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Dried Fecal Extract, Extracted Serum and Plasma, Tissue Culture Medium, Urine
Sample Volume 50 μL
Sensitivity 9.92 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Testosterone ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.